Learn More
Specifications
Specifications
| Content And Storage | 4°C. Do not freeze. |
| Product Line | Novus |
| Quantity | 50 mL |
| For Use With (Application) | Preparation of Working Solutions 1. Solution Pepsin is Ready to Use and Does Not Require Any Preparation. Protocol Recommendations 1. Deparaffinize Tissue Sections and Wash Slides with Wash Buffer. 2. Remove Excess Buffer From Slide Without Letting The Tissue Dry. 3. Depending on The Size of The Tissue Section Add 1-2 Drops of Pepsin Solution. 4. Incubate Slide for 5-10 Minutes (at 37 C. 5. Wash Slide with Wash Buffer and Proceed with Immunostaining.) |
| Type | Pepsin Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
