Learn More
Invitrogen™ Peptide Histidine-Methionine Monoclonal Antibody (03)
Shop All Thermo Scientific Products

Description
ABS 020-03-02 detects PHM in human samples. ABS 020-03-02 has been successfully used in ELISA procedures. PHM cross-reacts with PACAP (approximately 12%; incomplete, non-parallel inhibition). NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Specifications
Specifications
| Antigen | Peptide Histidine-Methionine |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 03 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | VIP |
| Gene Accession No. | P01282 |
| Gene Alias | Intestinal peptide PHM-27; Intestinal peptide PHV-42; MGC13587; Peptide histidine methioninamide 27; Peptide histidine valine 42; PHM27; prepro-VIP; vasoactive intestinal peptide; Vasoactive intestinal polypeptide; VIP; VIP peptides |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.