Learn More
CPC Scientific H-Ile-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Asn-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PEPY005A
Encompass
Y2 receptor agonist, released from the body's gastrointestinal tract in proportion to caloric intake, that significantly decreases appetite and reduces food intake, and thus may act as a drug for the treatment of obesity. ONE-LETTER SEQUENCE: IKPEAPGEDASPEELNRYYASLRHYLNLVTRQRY-NH2MOLECULAR FORMULA: C180H279N53O54MOLECULAR WEIGHT:4049.52STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [123583-37-9], SYNONYMS: PYY (3-36) (human)Peptide YY (3-36) (human)RESEARCH AREA: GastrointestinalREFERENCES: D. Grandt et al., Regulatory Peptides, 40, 161(1992)
Catalog No. 50-211-0148
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.