Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Peptidoglycan Monoclonal Antibody (3F6B3 (10H6))
GREENER_CHOICE

Catalog No. PIMA516509 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516509 Supplier Invitrogen™ Supplier No. MA516509
  • $752.00
    $614.65 / Each of 1

    Save $137.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Treatment with strong acid (Gram positive bacteria) or with a detergent such as SDS (Gram-negative bacteria) may be necessary to expose the epitope.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Peptidoglycan
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone 3F6B3 (10H6)
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with no preservative
Gene Alias PG
Host Species Mouse
Immunogen This antibody was raised against insoluble peptidoglycan obtained by TCA-heat and ethanol extraction of Streptococcus mutans BHT cells.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.