Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Pepto-Bismol™ Symptom-Relief Tablets

Catalog No. 19090828 Shop All Medique Products
Change view
Click to view available options
Quantity:
48/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-828 48/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-090-828 Supplier Medique Supplier No. 47367
Add to Cart
Add to Cart

For upset stomach, diarrhea, heartburn, and indigestion.

  • Each tablet contains: Subsalicylate 262mg

Specifications

Product Type Symptom Relief Tablets
Active Ingredients Subsalicylate 262 mg
For Use With (Application) For upset stomach, diarrhea, heartburn, indigestion
Format Chewable
Quantity 48/Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.