Learn More
United States Biological Corporation PEPTONE 2.5 KG
Supplier: United States Biological Corporation P33002.5KG
Peptone is an enzymatic digest of protein used to cultivate non-fastidious organisms, producing low-level growth. Deficient in carbohydrates, cystine and tryptophan. Appearance: Light tan, homogenous, free flowing powder pH: 7.0 �0.5 Amino Nitrogen: 6% Solubility (2%): Light yellow, clear, complete Total Nitrogen: 10% Reducing Sugars: Negative Undigested Protein: Negative Proteoses: Negative Nitrites: Negative Tryptophan: Negative Ash: 10% Moisture: 10% Typical Amino Acid Profile: Ala 8.5%, Arg 7.8%, Asp 6.2%, Cys 0.2%, Glu 9.8%, Gly 19.4%, His 0.8%, Ile 1.4%, Leu 2.9%, Lys 3.8%, Met 1.0%, Phe 1.9%, Pro 12.2%, Ser 3.7%, Thr 1.8%, Trp 0.05%, Tyr 0.7%, Val 2.2% Growth Supporting Properties (Agar/ Broth Formulations): Satisfactory Certificate of Origin: The raw animal products and enzymes, porcine tissue, used in the manufacturing of Peptone is derived from meat materials originating in the USA. During processing of the product, a batch is heated to a minimum of 80 �C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.