Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peptone, Powder, Beef, Spectrum™ Chemical
SDP

Catalog No. 18612174 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
5 kg
Packaging Type:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
18-612-174 100 g Poly Bottle
18-612-175 2.5 kg Poly Pail
18-612-176 5 kg Poly Pail
18-612-177 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-174 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PE117100GM
Add to Cart
Add to Cart

Specifications

Form Powder
Grade Reagent
Media Type Peptone
Product Type Media Additive
Packaging Type Poly Bottle
Quantity 100 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Do not feed to cattle or other ruminants. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.