Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 0.1 N Solution in Glacial Acetic Acid, (USP Volumetric Solution), Spectrum™ Chemical
SDP

Catalog No. 18612178 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-612-178 500 mL Amber Glass Bottle
18-612-179 1 L Amber Glass Bottle
18-612-180 2.5 L Amber Glass Bottle
18-612-181 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-612-178 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P107500ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7601-90-3 , 64-19-7 , 108-24-7

Specifications

Concentration 0.1 N ± 0.005 N
Packaging Amber Glass Bottle
Quantity 500 mL
Grade USP
Chemical Name or Material Perchloric Acid in Glacial Acetic Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.