Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 0.7, ULTREX™ II Ultrapure Reagent, J.T. Baker™

Catalog No. 02003551 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
PFA Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-551 500 mL PFA Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-551 Supplier Avantor Supplier No. 480601
Add to Cart
Add to Cart

Reagents for critical elemental analysis with less than 10 parts-per-trillion (ppt) levels of up to 65 elements

    For use in ICP (Inducutively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Packaging PFA Bottle
Specific Gravity 1L = 1.60kg
Quantity 500 mL
Formula Weight 100.46
Percent Purity 70%
Grade ULTREX™ II Ultrapure Reagent
Chemical Name or Material Perchloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.