Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 62.0%, MilliporeSigma™

Catalog No. 50948033 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-948-033 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-948-033 Supplier MilliporeSigma Supplier No. PX0396D4

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7601-90-3

Specifications

Packaging Glass Bottle, Pre-Packed
Assay Percent Range 60.0 to 62.0%
Quantity 2.5 L
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.