Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric acid concentrate, For 1L standard solution, 0.1 M HClO4 (0.1N), for titration in non-aqueous medium, Solstice

Catalog No. 60046057 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 Ea.
Packaging:
Ampoule
Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-057 6 x 1 Ea. Ampoule
60-011-53 1 L Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-057 Supplier Solstice Supplier No. 320466X1EA

missing translation for 'validMainframeMsg'

for 1L standard solution, 0.1 M HClO4 (0.1N), for titration in non-aqueous medium

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3 , 7732-18-5
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

CAS Min % ≥70.0000%, ≥20.0000%
CAS Max % <90.0000%, <30.0000%
Boiling Point 203°C
Density 1.66 g/cm3
Packaging Ampoule
Flash Point 113°C (235.4°F)
Linear Formula HClO4
Quantity 6 x 1 Ea.
UN Number UN1873
Physical Form Concentrate
Formula Weight 100.46
Chemical Name or Material Perchloric acid concentrate, For 1L standard solution, 0.1 M HClO4 (0.1N), for titration in non-aqueous medium
Show More Show Less
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375
Precautionary Statement H271-H290-H302-H314-H373
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 5.1; Packing Group:3: I; Proper Shipping Name: Perchloric acid
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.