Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric acid, Environmental Grade, 67-71%, Thermo Scientific™

Catalog No. AA38749K4
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38749K4 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA38749K4 Supplier Thermo Scientific Chemicals Supplier No. 38749K4

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Color Colorless
Melting Point -12°C
Boiling Point 203°C
Density 1.67 g/mL
Odor Odorless
Flash Point 113°C (235°F)
Quantity 2.5 L
Physical Form Liquid, Single Sub-Boiling Quartz Distillation
UN Number UN1873
Merck Index 14,7153
Formula Weight 100.46
Percent Purity 65 to 71%
Grade Environmental
Chemical Name or Material Perchloric acid
Show More Show Less
Hazard Category H271-H290-H302-H314-H335-H373
Hazard Statement GHS H Statement
H271-H314
May cause fire or explosion
strong oxidizer.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: PERCHLORIC ACID
EINECSNumber 231-512-4
RTECSNumber SC7500000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.