Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, in Glacial Acetic Acid, for Non-Aqueous Titrations, Certified, 0.1000N ±0.0005N, LabChem™

Catalog No. LC181001
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Amber Glass
HDPE Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC181001 500 mL Amber Glass
LC181002 1 L Amber Glass
LC181003 2.5 L HDPE Bottle
LC181004 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC181001 Supplier LabChem, Inc. Supplier No. LC181001
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-19-7 , 108-24-7 , 7601-90-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless to Yellow
Concentration 0.1000N ±0.0005N
Density 1.06g/mL
CAS Max % 97.04, 2.1, 0.86
Identification Passes Test
Odor Vinegar
Packaging Amber Glass
Specific Gravity 1.06g/mL
Traceability to NIST Traceable to NIST
Flash Point 40°C
Decomposition Information Carbon monoxide; Carbon dioxide; May release flammable gases; Thermal decomposition generates: Corrosive vapors
Quantity 500 mL
Physical Form Liquid
Linear Formula HClO4
UN Number UN2789
Solubility Information Soluble in water
Formula Weight 100.47
Grade Certified
Chemical Name or Material Perchloric Acid, in Glacial Acetic Acid, for Non-Aqueous Titrations
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.