Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid (TraceMetal™ Grade), Fisher Chemical™

Catalog No. A511500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
HDPE Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A511-500 500 mL Glass Bottle
A511-P500 500 mL HDPE Bottle
A511-P212 2.5 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A511-500
  • Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS.
  • Evaluated for 65 metals at < 1 ppb concentrations.
  • Manufactured, purified, and packaged in a Class1000 clean room.
  • Packaged in non-breakable polypropylene bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
  • Three year shelf life from time of manufacture - available on COA. 45 mm bottle neck size.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH 0.1
Color Colorless
Melting Point -18°C
Boiling Point 203°C
Density 1.660g/cm³
Packaging Glass Bottle
Certificate of Analysis Certificate of Lot Analysis included with each shipment.
Quantity 500 mL
Physical Form Liquid
UN Number 1873
Merck Index 15, 7267
Formula Weight 100.46
Percent Purity 65 to 71% (HClO4, w/w)
Grade TraceMetal
Chemical Name or Material Perchloric Acid
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
ShelfLife 3 years
DOTInformation DOT Class 5.1, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.