Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, TraceSELECT™, 67 to 72%, Solstice

Catalog No. 60047541 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-541 1 L Glass bottle
60-047-540 100 mL Glass bottle
60-047-542 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-047-541 Supplier Solstice Supplier No. 772271L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TraceSELECT™, for trace analysis, 65-71%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

pH >1
Color Colorless
Melting Point -18°C
Boiling Point 203°C
Density 1.66g/cm3 at 20°C
Odor Odorless
Packaging Glass bottle
Quantity 1 L
Physical Form Liquid
Linear Formula HClO4
UN Number UN1873
Formula Weight 100.46g/mol
Percent Purity 67 to 72%
Grade TraceSELECT
Chemical Name or Material Perchloric acid
Show More Show Less
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375
DOTInformation 8(5.1) / PGI
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.