Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD49d
Supplier:  Biolegend 304311
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD49d [9F10]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Common Marmoset, Cynomolgus, Dog (Canine), Horse (Equine), Rhesus, Sheep (Ovine), Squirrel Monkey; Apps: FC; Size: 25 tests
Catalog No. 50-166-119
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.