Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Mouse CD24 (M1/69) ….. Cat. No. 65-0242-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 650242U025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Mouse CD24 (M1/69) ….. Cat. No. 65-0242-U025 ..... Size 25 µgThe M1/69 antibody reacts with mouse CD24, a cell surface molecule also known as Heat Stable Antigen (HSA). CD24 is a GPI-linked glycoprotein involved in cell adhesion and is a ligand for CD62P (P-selectin). It is expressed on erthrocytes, thymocytes, peripheral lymphocytes and myeloid lineage cells. The expression of CD24 is used to delineate stages of of lymphocyte development in the bone marrow.
Catalog No. 50-210-3573
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.