Learn More
Biolegend PerCP/FireTM 780 anti-mouse
Supplier:  Biolegend 150646
Encompass_Preferred
PerCP/Fire™ 780 anti-mouse CD192 (CCR2) [SA203G11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-253-7981
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.