Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen peroxide solution, PERDROGEN™ 30% H2O2 H2O2 (w/w) puriss. p.a., Reag. ISO, Reag. Ph. Eur., Solstice

Catalog No. 6000807 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-008-07 1 L
60-008-08 500 mL
60-008-09 5 L
60-045-965 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-008-07 Supplier Solstice Supplier No. 316426X1L

PERDROGEN™ 30% H2O2 H2O2 (w/w) puriss. p.a., Reag. ISO, Reag. Ph. Eur.

Solstice ACS solvents for general laboratory use are:

  • a value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested tmeet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

UN Number 2014
Beilstein 3587191
Concentration 30% (w/w)
Formula Weight 34.01g/mol
Grade puriss. p.a.
Vapor Pressure 23.3mmHg (30°C)
Quantity 1 L
Chemical Name or Material Hydrogen Peroxide Solution, Reag. Ph. Eur. reag. ISO
Hazard Statement P280-P305 + P351 + P338 + P310
Precautionary Statement H318-H412
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.