Learn More
Honeywell™ Perfect Fit™ Aramid Fiber Seamless Knit Gloves
Catalog No. 19051329 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Large
Catalog No. Size
19-051-329 2X-Large
1 options
This item is not returnable. View return policy
Catalog No. 19-051-329 Supplier Honeywell™ Supplier No. KV18AJ10050
Add to Cart
Add to Cart
This item is not returnable. View return policy
Standard weight aramid fiber seamless gloves offer cut and abrasion protection.
Gloves do not provide thermal heat protection
  • Aramid fiber construction provides cut, slash and abrasion resistance
  • Construction provides comfortable fit and durability
  • No seams to rub or split
  • Can be laundered to extend service life
Specifications
ANSI Abrasion Rating ANSI 1
Construction Seamless Knit
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A3
Hand Ambidextrous
Latex Free Yes
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size 2X-Large
EN388 Standard EN 388
For Use With (Application) Automotive assembly, sheet metal, glass handling, metal stamping, metal fabrication, recycling, canning, injection moulding, paper/pulp industry
Model KV18AJ-100-50
Series Perfect Fit™
Yarn Gauge 7