Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Perfect Fit Aramid Fiber Sleeves

Catalog No. 19050008
Change view
Click to view available options
Length (English):
14 in.
Includes:
Thumbhole; 3 bar-tack for fingers
Without thumb hole
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Includes
19-050-008 14 in. Thumbhole; 3 bar-tack for fingers
19-816-531 14 in. Without thumb hole
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-050-008 Supplier Honeywell™ Supplier No. KVS214TH3BT
This item is not returnable. View return policy

Honeywell offers a wide range of knitted arm protection in various sizes and lengths. DuPont™ Kevlar™ engineered fiber helps prevent cuts and slashes and is inherently flame-resistant. Aramid sleeves offer an excellent choice for the glass and steel industries.

Perfect Fit - Kevlar™ Aramid fibers Sleeves feature DuPont™ Kevlar™ Aramid fibers brand fiber and are available in a variety of lengths, widths and configurations. Kevlar™ Aramid fibers engineered fiber is 5 times stronger than steel on an equal weight basis and provides excellent protection from cut, slash and abrasion hazards. Kevlar™ Aramid fibers is also flame resistant, and can withstand heat up to 900°F (400°C) without degrading.

TRUSTED_SUSTAINABILITY

Specifications

Thickness 2-ply
For Use With (Application) Metal sheet handling, recycling, automotive assembly, blade handling, knife sharpening, Glass handling
Includes Thumbhole; 3 bar-tack for fingers
Length (English) 14 in.
Length (Metric) 35.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.