Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfex™ 100% Synthetic Floor and Wall Washing Brush

Catalog No. 19042033B Shop All Perfex Corp. Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-042-033B Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-042-033B Supplier Perfex™ Supplier No. 3010R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Cleaning Brush
Color Red
For Use With (Application) Scrub stubborn deposits from rough unfinished surfaces like concrete, brick, blacktop, etc.
Length (English) Bristle 2 in.
Length (English) Brush 10.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.