Learn More
Beckman Coulter PerFix-nc (no centrifuge assay Kit), 75 Tests, IVD-CE
Supplier: Beckman Coulter B31167
The PerFix-nc Kit (no centrifuge assay Kit) has been developed to enhance the signal-to-noise ratio of intracellular staining and simplify the workload necessary for the sample preparation. Accurate detection of both intracellular and extracellular epitopes are obtained.