Learn More
AdipoGen Perfluorhexane mixed isomers
Supplier: AdipoGen CDXP0500G100
Chemical. CAS 355-42-0. Formula C6F14. MW 338.04. Used as a building block or intermediate for synthesis perfluorohexane plays a vital role as a reaction medium, especially for photooxidation reactions. Perfluorohexane is used as a coolant, photosensitizer in fluorous biphasic singlet oxygenation and an inert reaction medium. It is used in the electronic cooling liquid and insulator. Further, it finds application as an ultrasound contrast agent.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.