Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro-1-iodohexane, 97%

Catalog No. AC269940050
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC269940050 5 g
AC269941000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC269940050 Supplier Thermo Scientific Chemicals Supplier No. 269940050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 355-43-1 | C6F13I | 445.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-43-1
Molecular Formula C6F13I
Molecular Weight (g/mol) 445.95
InChI Key BULLJMKUVKYZDJ-UHFFFAOYSA-N
Synonym perfluorohexyl iodide, perfluoro-1-iodohexane, 1-iodoperfluorohexane, perfluoro-n-hexyl iodide, tridecafluorohexyl iodide, unii-a2a17jug4r, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodo, tridecafluoro-1-iodohexane, a2a17jug4r, hexane, tridecafluoro-6-iodo
PubChem CID 67733
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro-6-iodohexane
SMILES C(C(C(C(F)(F)I)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point -45.0°C
Density 2.0630g/mL
Boiling Point 116.0°C to 119.0°C
Flash Point >115°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Refractive Index 1.3270 to 1.3300
Linear Formula CF3(CF2)5I
Quantity 5 g
Specific Gravity 2.063
Solubility Information Solubility in water: insoluble.
Formula Weight 445.95
Percent Purity 97%
Chemical Name or Material Perfluoro-1-iodohexane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 206-586-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.