Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro(2-methyl-3-oxahexanoyl) fluoride, 97%

Catalog No. AAL1648306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1648306 5 g
AAL1648314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1648306 Supplier Thermo Scientific Chemicals Supplier No. L1648306
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2062-98-8 | C6F12O2 | 332.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2062-98-8
Molecular Formula C6F12O2
Molecular Weight (g/mol) 332.045
MDL Number MFCD00054657
InChI Key BCLQALQSEBVVAD-UHFFFAOYSA-N
Synonym 2,3,3,3-tetrafluoro-2-perfluoropropoxy propanoyl fluoride, undecafluoro-2-methyl-3-oxahexanoyl fluoride, perfluoro 2-methyl-3-oxahexanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy propanoyl fluoride, perfluoro-2-methyl-3-oxahexanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy, hfpo dimer, acmc-1cljd
PubChem CID 102740
IUPAC Name 2,3,3,3-tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoyl fluoride
SMILES C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)F

Specifications

Boiling Point 54°C to 56°C
Flash Point None
Refractive Index 1.3
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 332.06
Percent Purity 97%
Chemical Name or Material Perfluoro(2-methyl-3-oxahexanoyl) fluoride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 218-173-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.