Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents

Catalog No. PI67363
Encompass
Change view
Click to view available options
Standard Components:
Heptafluoro-butanoic acid anhydride
Pentafluoro-propanoic acid anhydride
Trifluoroacetic acid anhydride
Quantity:
25 g
100 g
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Components Quantity
PI67363 Trifluoroacetic acid anhydride 100 g
PI63164 Heptafluoro-butanoic acid anhydride 10 x 1 mL
PI63162 Heptafluoro-butanoic acid anhydride 100 g
PI65192 Pentafluoro-propanoic acid anhydride 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI67363 Supplier Thermo Scientific™ Supplier No. TS67363
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).

Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.

  • Used to prepare fluoracyl derivatives for GC/MS
  • Produce stable volatile derivatives for FID and ECD techniques
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC/MS
Standard Components Trifluoroacetic acid anhydride
Quantity 100 g
Storage Room Temperature
Hazards Corrosive, Flammable
Standard Concentration 98 to 100%

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.