Learn More
Sigma Organic Chemistry perfluoro benzyltetrali | 1G | MFCD00084640
Supplier: Sigma Organic Chemistry CDS0032731G

perfluoro benzyltetrali , 1G
About this Item:
MDL #: MFCD00084640
Molecular Weight: 774.13
UNSPSC Code: 12352200
Chemical Formula: C17F30
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.