Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorodecanoic acid, 97%

Catalog No. aab2229206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2229206 5 g
AAB2229214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AAB2229206 Supplier Thermo Scientific Chemicals Supplier No. B2229206
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 335-76-2 | C10HF19O2 | 514.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335-76-2
Molecular Formula C10HF19O2
Molecular Weight (g/mol) 514.09
MDL Number MFCD00004175
InChI Key PCIUEQPBYFRTEM-UHFFFAOYSA-N
Synonym perfluorodecanoic acid, nonadecafluorodecanoic acid, ndfda, pfda, perfluoro-n-decanoic acid, nonadecafluoro-n-decanoic acid, decanoic acid, nonadecafluoro, ccris 4417, decanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluoro, perfluorocapric acid
PubChem CID 9555
ChEBI CHEBI:35546
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 77°C to 82°C
Boiling Point 218°C
Linear Formula CF3(CF2)8CO2H
Quantity 5 g
UN Number UN2923
Beilstein 1810811
Formula Weight 514.09
Percent Purity 97%
Chemical Name or Material Perfluorodecanoic acid
Hazard Category H351-H360Df-H362
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 206-400-3
RTECSNumber HD9900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.