Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorododecane, 97%, Thermo Scientific™

Catalog No. AAL1682703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1682703 1 g
AAL1682706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1682703 Supplier Thermo Scientific Chemicals Supplier No. L1682703

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-59-5
Molecular Formula C12F26
Molecular Weight (g/mol) 638.09
MDL Number MFCD00042084
InChI Key WNZGTRLARPEMIG-UHFFFAOYSA-N
Synonym perfluorododecane, hexacosafluorododecane, n-perfluorododecane, perfluoro-n-dodecane, acmc-20ajy2
PubChem CID 78973
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-hexacosafluorododecane
SMILES C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Melting Point 72°C to 76°C
Density 1.5
Boiling Point 178°C
Linear Formula CF3(CF2)10CF3
Quantity 1 g
Beilstein 1811139
Formula Weight 638.09
Percent Purity 97%
Chemical Name or Material Perfluorododecane
EINECSNumber 206-204-8
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.