Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoroheptanoic acid, 98+%

Catalog No. AAA1209206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1209206 5 g
AAA1209214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1209206 Supplier Thermo Scientific Chemicals Supplier No. A1209206

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

CAS: 375-85-9 | C7HF13O2 | 364.062 g/mol

Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-85-9
Molecular Formula C7HF13O2
Molecular Weight (g/mol) 364.062
MDL Number MFCD00039604
InChI Key ZWBAMYVPMDSJGQ-UHFFFAOYSA-N
Synonym perfluoroheptanoic acid, tridecafluoroheptanoic acid, perfluoroenanthic acid, perfluoro-n-heptanoic acid, heptanoic acid, tridecafluoro, tridecafluoro-1-heptanoic acid, heptanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoro, n-perfluoroheptanoic acid, tridecafluoroenanthic acid, acmc-1agq8
PubChem CID 67818
ChEBI CHEBI:35547
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecafluoroheptanoic acid
SMILES C(=O)(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 31°C to 36°C (literature)
Density 1.792
Boiling Point 89°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.3 00
Quantity 5 g
UN Number UN3261
Beilstein 1808210
Solubility Information Insoluble in water.
Formula Weight 364.06
Percent Purity ≥98%
Chemical Name or Material Perfluoroheptanoic acid
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 206-798-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.