Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoropentanoic acid, 97%

Catalog No. AAB2156706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2156706 5 g
AAB2156714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2156706 Supplier Thermo Scientific Chemicals Supplier No. B2156706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 2706-90-3 | C5HF9O2 | 264.05 g/mol

It is used in the analysis of perfluorooctanesulfonate and related fluorochemicals in water and biological tissue samples by liquid chromatography-ion trap mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the analysis of perfluorooctanesulfonate and related fluorochemicals in water and biological tissue samples by liquid chromatography-ion trap mass spectrometry.

Solubility
Partly soluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2706-90-3
Molecular Formula C5HF9O2
Molecular Weight (g/mol) 264.05
MDL Number MFCD00040211
InChI Key CXZGQIAOTKWCDB-UHFFFAOYSA-N
Synonym perfluoropentanoic acid, perfluorovaleric acid, nonafluoropentanoic acid, nonafluorovaleric acid, pentanoic acid, nonafluoro, nonafluoro-1-pentanoic acid, valeric acid, nonafluoro, n-perfluoropentanoic acid, perfluoro-n-pentanoic acid, 2,2,3,3,4,4,5,5,5-nonafluoro-pentanoic acid
PubChem CID 75921
ChEBI CHEBI:83491
IUPAC Name 2,2,3,3,4,4,5,5,5-nonafluoropentanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Density 1.713
Boiling Point 140°C
Refractive Index 1.294
Quantity 5 g
UN Number UN3265
Beilstein 1800087
Solubility Information Partly soluble in water.
Formula Weight 264.06
Percent Purity 97%
Chemical Name or Material Perfluoropentanoic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 220-300-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.