Learn More
Cayman Chemical PerfluoropentnoIc AcId 25g
SDP
Supplier:  Cayman Chemical 3725425 G
Encompass_Preferred
An NSAID inhibits PGE2 and LTC4 production in A23187-stimulated isolated mouse peritoneal macrophages at 100 UM inhibits serum-induced chemotaxis of isolated rat peritoneal leukocytes at 25 and 50 Ug/ml induces hemolysis in isolated human erythrocytes and histamine release in isolated rat peritoneal mast cells in a light-dependent manner at 10 UM reduces carrageenan-induced paw edema and yeast-induced fever in rats and acetic acid-induced writhing in mice
Catalog No. 50-291-9450
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.