Learn More
Diagnostic Biosystems PERFORIN Perforin
Supplier:  Diagnostic Biosystems MOB55505
Encompass_Preferred
This antibody is specific to a 70 kDa protein, which is a potent cytolytic poreforming protein. Perforin is a specific marker of functionally active cytotoxic T-lymphocytes and natural killer cells. Perforin mediates cytolysis of target cells by membrane damage and apoptosis.
Catalog No. 50-238-5878
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.