Learn More
BioVendor, LLC Perforin Human ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC RGP021R
Encompass
Perforin Human ELISA, 96 wells. Research use only. Immunoassay kit to quantify human perforin in cell culture supernatants or buffered solution. Serum/plasma note: high & non-linear detection level is found in serum/plasma samples; high signal appears not dependent to the Ab pairs but probably to matrix and/or Perforin interaction with other molecules. Serum/plasma quantification is under the scientists responsibility and specific investigations. Calibration range: 62.52000 pg/ml. Detection limit: < 40 pg/ml. Synonyms: Perforin-1, P1, Cytolysin, PFP. Summary: Perforin has structural and functional similarities to C9. Like C9, Perforin creates transmembrane tubules and is capable of lysing non-specifically a variety of target cells. It is one of the main cytolytic proteins of cytolytic granules, and is known to be a key effector for T-cell- and NK-cell-mediated cytolysis. Defects in this gene cause familial HPLH2, a rare and lethal autosomal recessive disorder of early childhood.
Catalog No. 50-206-9232
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.