Learn More
HexArmor™ Chrome Series™ 4026 Mechanic Style Gloves
Catalog No. 19161981 Shop All Performance Fabrics, Inc Products
Click to view available options
Size:
2X-Large, 11
3X-Large, 12
Large, 9
Medium, 8
Small, 7
X-Large, 10
X-Small, 6
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-981 X-Small, 6
19-161-982 Small, 7
19-161-983 Medium, 8
19-161-984 Large, 9
19-161-985 X-Large, 10
19-161-986 2X-Large, 11
19-161-987 3X-Large, 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-161-981 Supplier HexArmor™ Supplier No. 4026XS (6)
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
The 4026 series mechanic style gloves offer comfort and cut level A8 resistance.
  • SuperFabric™ brand material palm provides ANSI/ISEA level A8 cut resistance
  • Back-of-hand impact protection system utilizes an advanced design to dissipate forceful blows
  • ANSI/ISEA 138 Level 1 impact protection
  • Mud Grip™ palm provides reliable grip in dry or light oil situations while increasing abrasion resistance
  • Interior seams implement a double stitched core-spun thread, adding further durability and longevity
  • Hi-vis color scheme increases visual awareness
  • Launderable
  • EN407 level 1 contact heat rated
Specifications
ANSI Abrasion Rating ANSI 2
Certifications/Compliance ANSI/ISEA 138 Level 1 impact protection
Certifications/Compliance (Europe) EN 407
Color Fluorescent Yellow, Black
Cut Resistance Rating ANSI A8
Material SuperFabric
Product Type Mechanic's Gloves
Size X-Small, 6
EN388 Standard EN 388:4X21FP
Model 4026-XS
Series Chrome Series™ 4026