Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HexArmor™ Chrome Series™ 4018 Mechanics Gloves

Catalog No. 19161842A Shop All Performance Fabrics, Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-842A Small
19-161-842D X-Large
19-161-842E 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-161-842A Supplier HexArmor™ Supplier No. 4018S 7
Add to Cart
Add to Cart
This item is not returnable. View return policy

SuperFabric™ material on the palm and knuckle patch provides ANSI Cut Level A6 protection. Synthetic leather on the palm increases abrasion resistance, and enhances grip capabilities in dry or light oil applications.

  • SuperFabric™ brand material on the palm and knuckle patch provides ANSI Cut Level A6 protection
  • Synthetic leather palm enhances grip in dry or light oil situations, while also increasing abrasion resistance
  • Interior seams implement a double stitched core-spun thread, adding further durability and longevity
  • Velcro™ wrist closure ensures a secure fit and maximizes comfort
  • Touchscreen capabilities
  • Machine washable

Specifications

Coating Material Uncoated
Color Black, Blue
Cut Resistance Rating ANSI A6
Hand Specific
Material Neoprene
Product Type Mechanic's Gloves
Size Small
For Use With (Application) Automotive Assembly/Repair
Glove Cuff Style Hook and Loop
Liner Material SuperFabric™
Palm Type Synthetic Leather
Series Chrome Series™ 4018
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.