Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Performance Plastic Chemical Wash Bottles with 1 Color Label
SCI_ED

Catalog No. S27213 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Soap
Sodium Hypochlorite (Bleach)
Water
Quantity:
1/Pk.
2/Pk.
6/Pk.
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Chemical Label Quantity
S27213 1000 mL Ethanol 6/Pk.
S27148 500 mL Acetone 1/Pk.
S15096 500 mL Soap 1/Pk.
S27149 500 mL Acetone 2/Pk.
S15097 500 mL Water 1/Pk.
S27157 1000 mL Acetone 1/Pk.
S27158 1000 mL Acetone 2/Pk.
S27159 1000 mL Acetone 6/Pk.
S27166 500 mL Distilled Water 1/Pk.
S27167 500 mL Distilled Water 2/Pk.
S27168 500 mL Distilled Water 6/Pk.
S27175 1000 mL Distilled Water 1/Pk.
S27176 1000 mL Distilled Water 2/Pk.
S27177 1000 mL Distilled Water 6/Pk.
S27184 500 mL Isopropanol 1/Pk.
S27185 500 mL Isopropanol 2/Pk.
S27186 500 mL Isopropanol 6/Pk.
S27193 1000 mL Isopropanol 1/Pk.
S27194 1000 mL Isopropanol 2/Pk.
S27195 1000 mL Isopropanol 6/Pk.
S27202 500 mL Ethanol 1/Pk.
S27203 500 mL Ethanol 2/Pk.
S27204 500 mL Ethanol 6/Pk.
S27211 1000 mL Ethanol 1/Pk.
S27212 1000 mL Ethanol 2/Pk.
S27220 500 mL Sodium Hypochlorite (Bleach) 1/Pk.
S27221 500 mL Sodium Hypochlorite (Bleach) 2/Pk.
S27222 500 mL Sodium Hypochlorite (Bleach) 6/Pk.
S27229 1000 mL Sodium Hypochlorite (Bleach) 1/Pk.
S27230 1000 mL Sodium Hypochlorite (Bleach) 2/Pk.
S27231 1000 mL Sodium Hypochlorite (Bleach) 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Catalog No. S27213 Supplier Eisco™ Supplier No. CHWB1040PK6
Only null left
Add to Cart
Add to Cart

Performance Plastic Wash Bottles are made of low-density polyethylene (LDPE) for superior quality, strength, and flexibility. Chemical labels feature the chemical information as well as hazard, safety, and first aid precautionary statements.

  • Chemical specific wash bottles with color coded caps and labels
  • Performance plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Easy to read labeling, including chemical specific information and safety warnings
  • Self-venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won't fade, crack, or peel
  • Sodium Hypochlorite (bleach) wash bottles are opaque (white) to protect contents from UV light rays
  • Other bottles are clear/transparent
  • Bottle labels comply with the Globally Harmonized System of Classification and Labeling of Chemicals (GHS)
  • Bottle Labels also comply with OSHA's HazCom 2012 requirements and Hazard Communication Standard 29 CFR 1910.1200

Specifications

Capacity (Metric) 1000 mL
Capacity (English) 202.88 oz.
Color Clear
Description One color
Includes Built in nozzle
Material LDPE
No. per Pack 6
Quantity 6/Pk.
Sterility Non-sterile
Certifications Hazard Communication Standard 29 CFR 1910.1200
Closure Color White
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Wide
Shape Cylindrical
Product Type Wash Bottle
For Use With (Equipment) Ethanol
Chemical Label Ethanol
Closure Material Plastic
Closure Size 6.5 cm
Closure Type Screw Cap
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.