Learn More
Syngene™ Performance Qualtification Documentation for Syngene Gel Imaging systems
GREENER_CHOICESDP
Catalog No. 01257293 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Syngene Gel Documentation Systems - Gbox and Gbox Mini
Catalog No. For Use With (Equipment)
01-257-293 Syngene Gel Documentation Systems - Gbox and Gbox Mini
1 options
Catalog No. 01-257-293 Supplier Syngene™ Supplier No. PQGEL
Add to Cart
Add to Cart
Syngene™ Performance Qualtification Documentation for Syngene Gel Imaging systems with synthetic gel and Kodak Step Wedge is for validation of Syngene Gbox Gel Doc systems
Specifications
Product Type Document
Quantity 1
For Use With (Equipment) Syngene Gel Documentation Systems - Gbox and Gbox Mini
WARNING: Cancer - www.P65Warnings.ca.gov