Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Peridotite Specimen (Igneous Rock)
SCI_ED

Catalog No. S24765 Shop All Eisco Products
Change view
Click to view available options
Quantity:
12/Pk.
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S24765 12/Pk.
S24764 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S24765 Supplier Eisco™ Supplier No. ESNG0047PK12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in comparing with other igneous rocks, particularly as part of lessons on Bowen's reaction series. EISCO Peridotite Specimen (Igneous Rock) is iron-rich ultramafic rock, with chemistry that suggests genesis from point deep within crust.

  • Useful as classroom demonstration of what is believed to make up most of earth's mantle, far below crust
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Contains abundant heavy elements such as iron and magnesium
  • Large crystals indicate slow cooling in magma chamber
  • Comes in clear labeled and barcoded bag for storage

Specifications

Quantity 12/Pk.
For Use With (Application) Comparing with other igneous rocks, particularly as part of lessons on Bowen's reaction series
Product Type Peridotite Study Specimen (Igneous Rock)
Width (English) Approx. 1 in.
Width (Metric) Approx. 3 cm
Packaging Clear labeled and barcoded bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.