Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Perilipin 1 Polyclonal Antibody, Alexa Fluor™ 647
GREENER_CHOICE

Catalog No. PIPA11051A6 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA11051A6 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA11051A6 Supplier Invitrogen™ Supplier No. PA11051A647
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-1051 detects perilipin A from rat and 3T3-L1 cell extract. The PA1-1051 immunogen is a synthetic peptide corresponding to the residues C E(502) P I L G R T Q Y S Q L R K K S(517) of rat Perilipin A. This sequence is 93% conserved in humans. The PA1-1051 immunizing peptide (Cat. # PEP-169) is available for use in neutralization and control experiments.

Adipose tissue is an energy reserve in animals and is strictly regulated in nondomestic species. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), resistin, and perilipin. Perilipin is an intracellular neutral lipid droplet protein that is hormonally regulated. This protein is localized exclusively to the surface of lipid droplets. In response to lypotic stimuli, perilipin is phosphorylated by protein kinase A. Once activated, perilipin has inhibitory affects upon hormone-sensitive lipase (HSL), a protein that mediates the hydrolysis of triacylglycerol, the major form of stored energy in the body. Perilipin expression is limited to adipocytes and steroidogenic cells. There are currently two known isoforms, Perilipin A and B. Both of these proteins are encoded by a single-copy gene and are the result of differential splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Perilipin 1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Alexa Fluor 647
Formulation PBS with 1.0mg/mL BSA and 0.05% sodium azide
Gene Plin1
Gene Accession No. P43884, Q8CGN5
Gene Alias 6030432J05Rik; adipocyte lipid droplet binding protein; FPLD4; Lipid droplet-associated protein; PERI; PERIA; perilipin; perilipin 1; perilipin A; perilipin B; perilipin-1; PLIN; PLIN1
Gene Symbols Plin1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the residues C E(502) P I L G R T Q Y S Q L R K K S(517) of rat Perilipin A.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103968, 25629
Target Species Mouse, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.