Learn More
Carbosynth LLC. Guinea Pig polyclonal Perilipin antibody
SDP
Supplier:  Carbosynth LLC. 20RPP004
Encompass_Preferred
Perilipins build a family of phosphoproteins. The predominant forms in adipocytes, perilipin A and B arise by alternative RNA splicing from a single gene, generating polypeptides of 57 and 46 kDa, respectively. The N-terminus, however, remains unchanged. The antiserum reacts specifically with perilipins (A and B) located at the surface of intracellular storage lipid droplets present e.g. in the adrenal gland, adipocytes of white and brown adipose tissue and cultured cells such as 3T3-L1 adipocytes and cultured steroidogenic adrenal cortical and Leydig cells.
Catalog No. 50-125-2899
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.