Learn More
Medchemexpress LLC HY-N7000 100mg Medchemexpress, Perillyl alcohol CAS:536-59-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN7000100MG
Encompass_Preferred
Medchemexpress, HY-N7000 100mg Perillyl alcohol CAS:536-59-4 Perillyl alcohol is a monoterpene isolated from the essential oils of lavendin, peppermint, spearmint, cherries, celery seeds, and several other plants. Perillyl alcohol is active in inducing apoptosis in tumor cells without affecting normal cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0489
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.