Learn More
Periodic Acid (White to Pale-Yellow Crystals or White Powder), Fisher BioReagents
Shop All Fisherbrand ProductsDescription
Specifications
Specifications
| pH | 1 |
| Color | White |
| Melting Point | 122°C |
| Density | 1.4 g/cm3 |
| Heavy Metals (as Pb) | 0.005% max. |
| Identification | Pass Test |
| Packaging | Poly Bottle |
| Assay Percent Range | ≥99 % |
| Quantity | 100 g |
| Physical Form | Solid |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Oxidizer: Contact with combustible/organic material may cause fire. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting. NFPA Health:3 Flammability:0 Instability:2 |
| DOTInformation | DOT Class 5.1, : Oxidizer |
| Recommended Storage | RT |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.