Learn More
Santa Cruz Biotechnology PERIOSTIN ANTIBODY (F-10)
Supplier:  Santa Cruz Biotechnology SC398631
Encompass
200 µg/mlAnti-Periostin Antibody (F-10) is a mouse monoclonal IgG1 κ Periostin antibody, cited in 13 publications, provided at 200 µg/mlraised against amino acids 537-836 mapping at the C-terminus of Periostin of human originAnti-Periostin Antibody (F-10) is recommended for detection of Periostin of mouse, rat and human origin by WB, IP, IF, IHC(P) and ELISA
Catalog No. NC1916795
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.