Learn More
StemExpress Peripheral Blood Monocyte-Derived Macrophages, Frozen, 1.5 million/vial
SDP
Supplier:  StemExpress PBMAC001.5C
Encompass
The isolation of mature macrophages from tissue can be difficult, therefore the use of monocyte-derived macrophages (MDM) for functional studies is a very attractive alternative. Non-monocytes are depleted from the mononuclear cell population using immunomagnetic particles leaving purified, untouched CD14+ monocytes. The untouched monocytes are cultured for 4-5 days with 10% FBS in the presence of M-CSF and IL-4. After culture, cells are checked for the expression of CD11b, CD18, CD68, and HLA-DR by flow cytometry.
Catalog No. 50-212-6329
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.