Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peripheral Node Addressin (PNAd) Rat anti-Mouse, Human, Clone: MECA-79, MilliporeSigma™

Catalog No. MABF2050MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2050MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2050MI Supplier MilliporeSigma Supplier No. MABF2050
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This rat monoclonal Anti-Peripheral Node Addressin (PNAd) Antibody, clone MECA-79 is tested in Flow Cytometry, Immunohistochemistry (Paraffin), , Immunoprecipitation, Inhibition, and Western Blotting for the detection of Peripheral node addressin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peripheral Node Addressin (PNAd)
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MECA-79
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Rat
Immunogen Collagenase-dispersed lymph node stromal cells from BALB/c mice.
Purification Method Purified by ion-exchange chromatography
Quantity 200 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone MECA-79 detects Peripheral Node Addressin (PNAd) in human and mouse.
Target Species Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.