Learn More
Agilent BioTek Peristaltic Pump 1536-well Dispenser Cassette with Plastic Tips
Shop All Agilent Technologies ProductsDescription
- 1 μL increments
- 0.01 in. diameter plastic tips
Specifications
Specifications
| Capacity (Metric) | 500 nL to 50 μL |
| For Use With (Equipment) | EL406 Microplate Washer, 406 FX Microplate Washer, Multiflo FX, 96- and 384-well Dispensing |
| Product Type | Low Volume 1536 Well Cassette |