Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PERK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582537 Supplier Invitrogen™ Supplier No. PA582537
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NAWLEAPPEK WQEKMDEIWL KDESTDWPLS SPSPMDAPSV KIRRMDPFAT KEHIEIIAPS PQRSRSFSVG ISCDQTSSSE SQFSPLEFSG MDHEDISESV DAAYNLQDSC LTDCDVEDGT MDGNDE Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 85%.

EIF2AK3 (PERK) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER. Defects in EIF2AK3 are the cause of Wolcott-Rallison syndrome (WRS), also known as multiple epiphyseal dysplasia with early-onset diabetes mellitus. WRS is a rare autosomal recessive disorder, characterized by permanent neonatal or early infancy insulin-dependent diabetes and, at a later age, epiphyseal dysplasia, osteoporosis, growth retardation and other multisystem manifestations, such as hepatic and renal dysfunctions, mental retardation and cardiovascular abnormalities.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PERK
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EIF2AK3
Gene Accession No. Q9NZJ5
Gene Alias 2.7.11.1; AI427929; EIF2AK3; eukaryotic translation initiation factor 2 alpha kinase 3; eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; Pancreatic eIF2-alpha kinase; PEK; Perk; PRKR-like endoplasmic reticulum kinase; WRS
Gene Symbols EIF2AK3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EIF2AK3. Recombinant protein control fragment (Product #RP-90474).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9451
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.