Learn More
Description
MA1-06821 detects large heparan sulphate proteoglycan in human, mouse, bovine, cod, wolffish, and rat samples. MA1-06821 has sucessfully been used in Western blotting, immunocytochemistry, immunoprecipitation, and immunohistochemistry procedures. By Western blot it detects a >400 kDa band representing large HSPG. The MA1-06821 immunogen is a high molecular mass material derived from the Engelbreth-Holm-Swarm (EHS) tumor matrix containing laminin, entactin and HSPG.
Specifications
Specifications
| Antigen | Perlecan |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | A7L6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | HSPG2 |
| Gene Accession No. | P98160, Q05793 |
| Gene Alias | AI852380; Basement membrane-specific heparan sulfate proteoglycan core protein; Endorepellin; endorepellin (domain V region); heparan sulfate proteoglycan 2; HSPG; Hspg2; LG3 peptide; LOC313641; Pcn; per; perlecan; perlecan (heparan sulfate proteoglycan 2); perlecan proteoglycan; PLC; PRCAN; RP11-132G19.2; SJA; SJS; SJS1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
